Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E/Z)-ZINC09659342 | 591726-26-0 | 99.7% | C23H15F3N2O4 | 10MG
SDP

Catalog No. 5000258306
Encompass_Preferred

Small-molecule inhibitor of the Lbc-RhoA protein-protein interaction for use in biochemical and cell-based research. Supplied as a solid with high reported purity, it is commonly prepared as DMSO stock solutions for in vitro assays. For research use only; not for human or therapeutic use.

  • High reported purity (99.7%).
  • Molecular formula C23H15F3N2O4.
  • Molecular weight 440.37 g·mol⁻¹.
  • Supplied as a solid suitable for solution preparation.
  • Active in assays targeting RhoA signaling pathways.
  • Supplied with analytical data and purity documentation.

Catalog No. 50-002-58306 Supplier Medchemexpress LLC Supplier No. HY145915A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Small-molecule inhibitor of the Lbc-RhoA protein-protein interaction for use in biochemical and cell-based research. Supplied as a solid with high reported purity, it is commonly prepared as DMSO stock solutions for in vitro assays. For research use only; not for human or therapeutic use.

  • High reported purity (99.7%).
  • Molecular formula C23H15F3N2O4.
  • Molecular weight 440.37 g·mol⁻¹.
  • Supplied as a solid suitable for solution preparation.
  • Active in assays targeting RhoA signaling pathways.
  • Supplied with analytical data and purity documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.