Learn More
TARGETMOL CHEMICALS INC E Z-Zotiraciclib 10MG
SDP
Catalog No. 502875998
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (E/Z)-Zotiraciclib ((E/Z)-TG02) effectively inhibits CDK2, JAK2, and FLT3 with IC50s of 13 nM, 73 nM, and 56 nM, respectively. Purity 99.12%
Catalog No. 50-287-5998 Supplier TARGETMOL CHEMICALS INC Supplier No. T2150310MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. (E/Z)-Zotiraciclib ((E/Z)-TG02) effectively inhibits CDK2, JAK2, and FLT3 with IC50s of 13 nM, 73 nM, and 56 nM, respectively. Purity 99.12%