Learn More
Medchemexpress LLC HY-15166 5mg , (E/Z)-Zotiraciclib CAS:937270-47-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY151665MG
Encompass_Preferred
HY-15166 5mg (E/Z)-Zotiraciclib CAS: 937270-47-8 (E/Z)-Zotiraciclib ((E/Z)-TG02) is a potent inhibitor of CDK2, JAK2, and FLT3. (E/Z)-Zotiraciclib ((E/Z)-TG02) can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9194