Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E/Z)-zotiraciclib citrate | 1204918-73-9 | 99.8% | 564.59 g/mol | C29H32N4O8 | 10 MG
SDP

Catalog No. 5000287189
Encompass_Preferred

(E/Z)-Zotiraciclib citrate is the citrate salt form of zotiraciclib, a small-molecule kinase inhibitor active against CDK2, JAK2, and FLT3. It is supplied as an off-white to light yellow solid for laboratory research use, and is characterized by high purity and defined storage guidance for both solid and solution forms.

  • Potent inhibitor of CDK2, JAK2, and FLT3, suitable for kinase research.
  • Citrate salt form with molecular weight 564.59 g/mol.
  • Chemical formula C29H32N4O8.
  • High purity (≈99.8%), appropriate for biochemical and cell-based assays.
  • Solid physical form: off-white to light yellow powder.
  • Recommended storage sealed, away from moisture; freeze solutions for long-term storage.
  • Offered in multiple pack sizes, including small milligram quantities and solution formats for convenience.

Catalog No. 50-002-87189 Supplier Medchemexpress LLC Supplier No. HY15166B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-Zotiraciclib citrate is the citrate salt form of zotiraciclib, a small-molecule kinase inhibitor active against CDK2, JAK2, and FLT3. It is supplied as an off-white to light yellow solid for laboratory research use, and is characterized by high purity and defined storage guidance for both solid and solution forms.

  • Potent inhibitor of CDK2, JAK2, and FLT3, suitable for kinase research.
  • Citrate salt form with molecular weight 564.59 g/mol.
  • Chemical formula C29H32N4O8.
  • High purity (≈99.8%), appropriate for biochemical and cell-based assays.
  • Solid physical form: off-white to light yellow powder.
  • Recommended storage sealed, away from moisture; freeze solutions for long-term storage.
  • Offered in multiple pack sizes, including small milligram quantities and solution formats for convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.