Learn More
Medchemexpress LLC (E/Z)-Zotiraciclib citrate | 1204918-73-9 | C29H32N4O8 | 10 MM 1 ML
SDP
Catalog No. 5000373998
Encompass_Preferred
(E/Z)-Zotiraciclib citrate is a potent inhibitor of CDK2, JAK2, and FLT3, intended for research use only. It appears as a solid with an off-white to light yellow color and has a high purity of 99.83%. It is soluble in DMSO at 250 mg/mL, with various in vivo dissolution protocols available.
  • Potent CDK2 inhibitor
  • Potent JAK2 inhibitor
  • Potent FLT3 inhibitor
  • For research use only
  • Off-white to light yellow solid appearance
  • High purity of 99.83%
Catalog No. 50-003-73998 Supplier Medchemexpress LLC Supplier No. HY15166B10MM/1ML
(E/Z)-Zotiraciclib citrate is a potent inhibitor of CDK2, JAK2, and FLT3, intended for research use only. It appears as a solid with an off-white to light yellow color and has a high purity of 99.83%. It is soluble in DMSO at 250 mg/mL, with various in vivo dissolution protocols available.
  • Potent CDK2 inhibitor
  • Potent JAK2 inhibitor
  • Potent FLT3 inhibitor
  • For research use only
  • Off-white to light yellow solid appearance
  • High purity of 99.83%