Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E/Z)-Zotiraciclib citrate | 1204918-73-9 | C29H32N4O8 | 100 MG
SDP

Catalog No. 5000374187
Encompass_Preferred

(E/Z)-Zotiraciclib citrate is a potent inhibitor targeting CDK2, JAK2, and FLT3. It is intended for research use only.

  • Potent CDK2, JAK2, and FLT3 inhibitor
  • Purity: 99.83%
  • Molecular weight: 564.59
  • Chemical formula: C29H32N4O8
  • CAS number: 1204918-73-9
  • Appearance: Solid, off-white to light yellow
  • Solubility in DMSO: 250 mg/mL

Catalog No. 50-003-74187 Supplier Medchemexpress LLC Supplier No. HY15166B100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-Zotiraciclib citrate is a potent inhibitor targeting CDK2, JAK2, and FLT3. It is intended for research use only.

  • Potent CDK2, JAK2, and FLT3 inhibitor
  • Purity: 99.83%
  • Molecular weight: 564.59
  • Chemical formula: C29H32N4O8
  • CAS number: 1204918-73-9
  • Appearance: Solid, off-white to light yellow
  • Solubility in DMSO: 250 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.