Learn More
Medchemexpress LLC E0924G | 2909474-29-7 | 99.2% | 232.24 | 1 ML
SDP
Catalog No. 5000390445
Encompass_Preferred
E0924G is an orally active activator for PPARδ with an EC50 of 2.82 μM. It promotes the upregulation of osteoprotegerin (OPG) with an EC50 of 0.29 μM. E0924G reduces RANKL-induced osteoclast differentiation and inhibits F-actin ring formation in RAW264.7 macrophages. It also regulates bone density and bone loss in ovariectomized (OVX) and age-related osteoporosis models.
  • Orally active activator for PPARδ
  • Promotes upregulation of osteoprotegerin (OPG)
  • Reduces RANKL-induced osteoclast differentiation
  • Inhibits F-actin ring formation in RAW264.7 macrophages
  • Regulates bone density and bone loss
Catalog No. 50-003-90445 Supplier Medchemexpress LLC Supplier No. HY16833610MM/1ML
E0924G is an orally active activator for PPARδ with an EC50 of 2.82 μM. It promotes the upregulation of osteoprotegerin (OPG) with an EC50 of 0.29 μM. E0924G reduces RANKL-induced osteoclast differentiation and inhibits F-actin ring formation in RAW264.7 macrophages. It also regulates bone density and bone loss in ovariectomized (OVX) and age-related osteoporosis models.
  • Orally active activator for PPARδ
  • Promotes upregulation of osteoprotegerin (OPG)
  • Reduces RANKL-induced osteoclast differentiation
  • Inhibits F-actin ring formation in RAW264.7 macrophages
  • Regulates bone density and bone loss