Learn More
Leica Biosystems E0SIN 1 GAL
Catalog No. NC1093844
Encompass
Eosin Y is an alcohol-based, ready-to-use secondary stain used primarily in histology for routine hematoxylin and eosin staining. It may also be used in cytology for fast cytoplasmic staining of rush specimens also using the H&E technique. Available in various quantities.
Catalog No. NC1093844 Supplier Leica Biosystems Supplier No. 3801600
Eosin Y is an alcohol-based, ready-to-use secondary stain used primarily in histology for routine hematoxylin and eosin staining. It may also be used in cytology for fast cytoplasmic staining of rush specimens also using the H&E technique. Available in various quantities.