Learn More
Pipette Supplies, Inc. E1 ClipTip Lithium-Ion Battery, Single and Multichannel (Thermo Scientific)
SDP
Catalog No. NC2943550
Encompass
E1 ClipTip Lithium-Ion Battery, Single and Multichannel (Thermo Scientific)Thermo Scientific OEM Rechargeable Lithium Ion Battery Assembly, for the E1 Clip Tip Single Channel and Multichannel pipettes.3.7V, 1230mAh, 4.55WhBattery must be charged prior to use. See user manual below for instruction.Made in ChinaPlease note that we only ship this battery via Ground Shipping.
Catalog No. NC2943550 Supplier Pipette Supplies, Inc. Supplier No. TS 2215640
E1 ClipTip Lithium-Ion Battery, Single and Multichannel (Thermo Scientific)Thermo Scientific OEM Rechargeable Lithium Ion Battery Assembly, for the E1 Clip Tip Single Channel and Multichannel pipettes.3.7V, 1230mAh, 4.55WhBattery must be charged prior to use. See user manual below for instruction.Made in ChinaPlease note that we only ship this battery via Ground Shipping.