Learn More
Enzo Life Sciences Ubiquitin activating enzyme E1 (human), (recombinant) (His-tag) (50 µg)
SDP
Catalog No. NC1275796
Encompass_Preferred
Essential ubiquitinylation pathway enzyme. MW: 118 kDa. Purity: ≥90% (SDS-PAGE). Formulation: In 50mM TRIS-hydrochloric acid, containing 1.0mM dithiothreitol. Source: Produced in E. coli BL21 (λDE3) expression system and containing a C-terminal His6-tag. Full length human ubiquitin-activating enzyme E1. UniProt: P22314. Handling: Avoid freeze/thaw cycles. Long Term Storage: -80°C
Catalog No. NC1275796 Supplier Enzo Life Sciences Supplier No. BMLUW94100050
Essential ubiquitinylation pathway enzyme. MW: 118 kDa. Purity: ≥90% (SDS-PAGE). Formulation: In 50mM TRIS-hydrochloric acid, containing 1.0mM dithiothreitol. Source: Produced in E. coli BL21 (λDE3) expression system and containing a C-terminal His6-tag. Full length human ubiquitin-activating enzyme E1. UniProt: P22314. Handling: Avoid freeze/thaw cycles. Long Term Storage: -80°C