Learn More
Malvern Panalytical E1 Protection Foil,25/Pk
Catalog No. NC2059775
Encompass
**Per supplier requirements this item has a minimum qty of 3 **Box with 25 pre-cut 6 um polypropylene protective foils to protect the optical path of the Epsilon 1 system against spillage of liquids and loose powders.
Catalog No. NC2059775 Supplier Malvern Panalytical Supplier No. 9430 500 07221
**Per supplier requirements this item has a minimum qty of 3 **Box with 25 pre-cut 6 um polypropylene protective foils to protect the optical path of the Epsilon 1 system against spillage of liquids and loose powders.