Learn More
Miele, Inc. E 118 Full Insert for 38 Petri dishes (Ø 100 mm)
Catalog No. NC1511418
Encompass_Preferred
E 118 Mesh Basket for the optimum loading of petri dishes.
Suitable for 38 half petri dishes with Ø 100 mm
Equipped with 38 holders, height 70 mm, distance approx. 26 mm
For use in upper basket
Visit www.mieleusa.com/professional for technical information.
Catalog No. NC1511418 Supplier Miele, Inc. Supplier No. 03830270
E 118 Mesh Basket for the optimum loading of petri dishes.
Suitable for 38 half petri dishes with Ø 100 mm
Equipped with 38 holders, height 70 mm, distance approx. 26 mm
For use in upper basket
Visit www.mieleusa.com/professional for technical information.