Learn More
Alkali Scientific E1200 myGel InstaView Gel Tank
SDP
Catalog No. 503029549
Encompass_Preferred
For labs that prefer to use a separate power supply the Alkali Scientific E1200 myGel InstaView Gel Tank w/ Blue LED Illuminator is the perfect choice for running viewing and documenting electrophoretic separations The Alkali Scientific E1200 myGel InstaView Gel Tank w/ Blue LED Illuminator accommodates one large (10 5x10cm) or one small (10 5x6cm) gel tray and is supplied with trays combs and a leak-proof casting stand The gel tank lid includes attached power leads which prevent the unit from operating without the lid properly in place Any standard power supply can be connected including the Alkali Scientific myVolt Mini and myVolt Touch The Alkali Scientific E1200 myGel InstaView Gel Tank w/ Blue LED Illuminator is shaped to allow the mini LED transilluminator (included) to slide underneath Blue light at a wavelength of 465nm provides excitation of common green fluorescent stains such as SmartGlow SYBR Green and GelGreen An orange filter is incorporated into the gel tank lid f
Catalog No. 50-302-9549 Supplier Alkali Scientific Supplier No. ES1200
For labs that prefer to use a separate power supply the Alkali Scientific E1200 myGel InstaView Gel Tank w/ Blue LED Illuminator is the perfect choice for running viewing and documenting electrophoretic separations The Alkali Scientific E1200 myGel InstaView Gel Tank w/ Blue LED Illuminator accommodates one large (10 5x10cm) or one small (10 5x6cm) gel tray and is supplied with trays combs and a leak-proof casting stand The gel tank lid includes attached power leads which prevent the unit from operating without the lid properly in place Any standard power supply can be connected including the Alkali Scientific myVolt Mini and myVolt Touch The Alkali Scientific E1200 myGel InstaView Gel Tank w/ Blue LED Illuminator is shaped to allow the mini LED transilluminator (included) to slide underneath Blue light at a wavelength of 465nm provides excitation of common green fluorescent stains such as SmartGlow SYBR Green and GelGreen An orange filter is incorporated into the gel tank lid f