Learn More
Medchemexpress LLC HY-18233 5mg Medchemexpress, E1210 CAS:936339-60-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY182335MG
Encompass_Preferred
Medchemexpress, HY-18233 5mg E1210 CAS:936339-60-5 E1210 is a first-in-class, broad-spectrum and orally active antifungal. E1210 has a mechanism of action-inhibition of fungal glycosylphosphatidylinositol (GPI) biosynthesis[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0810