Learn More
Miele, Inc. E 137 Full Insert for 56 Petri dishes (Ø 100 mm) (for use with E 136 only)
Catalog No. NC1511419
Encompass_Preferred
E 137 Mesh Basket for optimum loading of 56 half petri dishes.
Suitable for 56 half petri dishes with Ø 100 mm
Equipped with 56 holders, height 2 7/8 (70) mm, distance apart approx. 1 1/8 (26) mm
For use in lower basket
Visit www.mieleusa.com/professional for technical information.
Catalog No. NC1511419 Supplier Miele, Inc. Supplier No. 03830290
E 137 Mesh Basket for optimum loading of 56 half petri dishes.
Suitable for 56 half petri dishes with Ø 100 mm
Equipped with 56 holders, height 2 7/8 (70) mm, distance apart approx. 1 1/8 (26) mm
For use in lower basket
Visit www.mieleusa.com/professional for technical information.