Learn More
Grainger Hallowell Locker Finished End Panel, Steel, 72 in. Height, 12 in. Depth, 1 ea.
Catalog No. 502793163
Encompass_Preferred
This finished end panel provides a clean, aesthetic look for exposed row ends in locker installations. Designed for durability and universal compatibility, the panel is constructed from solid steel and features a powder-coated finish for enhanced wear resistance. It is intended for use with 72 in. high lockers with a depth of 12 in.
  • Constructed from strong and durable steel
  • Finished with a powder coating for longevity
  • Designed for 72 in. high lockers with 12 in. depth
  • Solid panel construction without interior perforations
  • Features a minimum of 30% post-consumer recycled content
  • GREENGUARD certified
  • Includes one universal end panel
Catalog No. 50-279-3163 Supplier Grainger Supplier No. 1AET2
This finished end panel provides a clean, aesthetic look for exposed row ends in locker installations. Designed for durability and universal compatibility, the panel is constructed from solid steel and features a powder-coated finish for enhanced wear resistance. It is intended for use with 72 in. high lockers with a depth of 12 in.
  • Constructed from strong and durable steel
  • Finished with a powder coating for longevity
  • Designed for 72 in. high lockers with 12 in. depth
  • Solid panel construction without interior perforations
  • Features a minimum of 30% post-consumer recycled content
  • GREENGUARD certified
  • Includes one universal end panel