Learn More
Medchemexpress LLC HY-116463 10mg Medchemexpress, E1R CAS:1301211-78-8 Purity:>98%
SDP
Catalog No. 501969418
Encompass_Preferred
Medchemexpress, HY-116463 10mg E1R CAS:1301211-78-8 E1R is a positive allosteric modulator of sigma-1 receptors (Sig1R PAM) with cognition-enhancing activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9418 Supplier Medchemexpress LLC Supplier No. HY11646310MG
Medchemexpress, HY-116463 10mg E1R CAS:1301211-78-8 E1R is a positive allosteric modulator of sigma-1 receptors (Sig1R PAM) with cognition-enhancing activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.