Learn More
Cayman Chemical (E)-2-(2-Chlorostyryl)-3,5,6-trimethylpyrazine, Purity: ≥98%, CAS: 1000672-89-8, 25 mg
SDP
Catalog No. 502896018
Encompass_Preferred
A stilbene derivative with antioxidant and anticancer activities; stimulates proliferation of hydrogen peroxide-damaged ECV-304 cells (EC50 = 24.9 nM); reduces hydrogen peroxide-induced release of LDH in and increases viability of HUVECs; reverses hydrogen peroxide-induced release of MDA and decreases in SOD and GSH-Px activities and increases cNOS activity and NO production in HUVECs; induces cell death of A549 NSCLC cells in an IRE1α-dependent manner
Catalog No. 50-289-6018 Supplier Cayman Chemical Supplier No. 900320225 MG
A stilbene derivative with antioxidant and anticancer activities; stimulates proliferation of hydrogen peroxide-damaged ECV-304 cells (EC50 = 24.9 nM); reduces hydrogen peroxide-induced release of LDH in and increases viability of HUVECs; reverses hydrogen peroxide-induced release of MDA and decreases in SOD and GSH-Px activities and increases cNOS activity and NO production in HUVECs; induces cell death of A549 NSCLC cells in an IRE1α-dependent manner