Learn More
MSC Betco E2 Antibacterial Foaming Skin Cleanser, 1000 mL, NSF E2 Certified, Fragrance-Free
Catalog No. 502837495
Encompass
The E2 Antibacterial Foaming Skin Cleanser is an NSF E2 International Certified, fragrance-free hand wash designed for repeated use. It contains 0.13% benzalkonium chloride to effectively reduce 99.99% of common germs within 15 seconds. This mild, clean-rinsing foam is ideal for the food service industry, leaving hands feeling clean without dryness or irritation, and encourages frequent hand washing.
  • NSF E2 International Certified and FDA compliant
  • Contains 0.13% benzalkonium chloride for >99.99% broad spectrum germ reduction in 15 seconds
  • Thick foam leaves hands feeling clean
  • Will not cause dryness or irritation
  • Encourages frequent hand washing due to mildness and clean rinsing
  • Leaves hands without stickiness, dryness, or residue
  • Sealed packaging keeps formula free of contaminants and provides 99% product evacuation
  • Recommended for repeated use
Catalog No. 50-283-7495 Supplier MSC Supplier No. 73070229
The E2 Antibacterial Foaming Skin Cleanser is an NSF E2 International Certified, fragrance-free hand wash designed for repeated use. It contains 0.13% benzalkonium chloride to effectively reduce 99.99% of common germs within 15 seconds. This mild, clean-rinsing foam is ideal for the food service industry, leaving hands feeling clean without dryness or irritation, and encourages frequent hand washing.
  • NSF E2 International Certified and FDA compliant
  • Contains 0.13% benzalkonium chloride for >99.99% broad spectrum germ reduction in 15 seconds
  • Thick foam leaves hands feeling clean
  • Will not cause dryness or irritation
  • Encourages frequent hand washing due to mildness and clean rinsing
  • Leaves hands without stickiness, dryness, or residue
  • Sealed packaging keeps formula free of contaminants and provides 99% product evacuation
  • Recommended for repeated use