Learn More
Mettler Toledo™ E2 OIML Class Standard Weight Set (E2 AL)
Catalog No. 01912672 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Weight: 1 mg to 1 kg
Catalog No.
01-912-672 Weight: 1 mg to 1 kg
1 options
Catalog No. 01-912-672 Supplier Mettler Toledo™ Supplier No. 158856
Add to Cart
Add to Cart
Request a Quote
E2 OIML class weight set is housed in an aluminum box and includes accessories for handling and cleaning.
  • Without calibration certificate
  • For Standard Enviroments: Affordable quality weights for regular routine testing without the need for traceability
  • High-grade stainless steel: Low magnetic permeability and high corrosion resistance ensure reliable values
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Weight
Material Stainless Steel
Case Material Aluminum
Weight (Metric) 1 mg to 1 kg
Density 8000 (± 60) kg/m3
Includes 25 Pieces; Without certificate