Learn More
Ubpbio E2 Screening Kit
Catalog No. 501965153
Encompass_Preferred
The E2 Screening Kit is useful for identification of the E2 Ub-conjugating enzyme(s) for a specific E3 Ub ligase and/or a protein substrate. The kits contains UbE1, Ub, 29 E2s, and reaction buffer.
Catalog No. 50-196-5153 Supplier Ubpbio Supplier No. J1100
The E2 Screening Kit is useful for identification of the E2 Ub-conjugating enzyme(s) for a specific E3 Ub ligase and/or a protein substrate. The kits contains UbE1, Ub, 29 E2s, and reaction buffer.