Learn More
Aviva Systems Biology Corp E2F1 Antibody - N-terminal region (P100821_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100821_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against E2F1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human E2F1. Alternate names: E2F-1, RBBP3, RBP3, RBAP1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62730