Learn More
Aviva Systems Biology Corp E2F2 Antibody - middle region (ARP31611_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31611_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against E2F2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human E2F2. Alternate names: E2F-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54283