Learn More
Aviva Systems Biology Corp E2f5 Antibody - C-terminal region (ARP57966_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57966_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against E2f5. It was validated on Western Blot. . Alternate names: AU024671, E2F-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-286-421