Learn More
Aviva Systems Biology Corp E2f7 Antibody - N-terminal region (ARP37583_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37583_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against E2f7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of mouse E2f7. Alternate names: A630014C11Rik, D10Ertd739eApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53080