Learn More
Electron Microscopy Sciences Edwards E2M1.5 Vacuum Pump, 110 V
SDP
Catalog No. 5025707
Encompass
This miniature two stage pump features an alternative inlet connection position at the side of the pump so that overall installation height can be reduced to a minimum when required. 1.3 cfm (190L/min) displacement and an ultimate vacuum without gas ballast of 1.1x10-3 torr. The max. inlet pressure for water vapor is 11.3 torr. Maxium water vapor pumping rate is 0.035 lb/hr.
Catalog No. 50-257-07 Supplier Electron Microscopy Sciences Supplier No. 91004
This miniature two stage pump features an alternative inlet connection position at the side of the pump so that overall installation height can be reduced to a minimum when required. 1.3 cfm (190L/min) displacement and an ultimate vacuum without gas ballast of 1.1x10-3 torr. The max. inlet pressure for water vapor is 11.3 torr. Maxium water vapor pumping rate is 0.035 lb/hr.