Learn More
MARSHALL SCIENTIFIC Edwards E2M1.5 Vacuum Pump
SDP
Supplier:  MARSHALL SCIENTIFIC EDA37132902
Encompass_Preferred
The EM series pumps are rugged mechanical oil-sealed pumps that are compact and quiet, The EM series also features advanced lubrication circuits, high reliability, and many different available accessories. These pumps are renowned for their high ultimate vacuum, rapid pumping speed, quiet operation, and ability to handle vapour. The EM series has become the industry standard for laboratory and light industrial applications. Ultimate vacuum of 0.003 mbar and peak pumping speed of 1.2 CFM.
Catalog No. NC2888787