Learn More
MARSHALL SCIENTIFIC Edwards E2M18 Vacuum Pump
SDP
Catalog No. NC2888789
Encompass_Preferred
The EM series pumps are rugged mechanical oil-sealed pumps that are compact and quiet, The EM series also features advanced lubrication circuits, high reliability, and many different available accessories. These pumps are renowned for their high ultimate vacuum, rapid pumping speed, quiet operation, and ability to handle vapour. The EM series has become the industry standard for laboratory and light industrial applications. Ultimate vacuum of 0.001 mbar and peak pumping speed of 12.1 CFM.
Catalog No. NC2888789 Supplier MARSHALL SCIENTIFIC Supplier No. EDA36317984
Add to Cart
Add to Cart
Request a Quote
The EM series pumps are rugged mechanical oil-sealed pumps that are compact and quiet, The EM series also features advanced lubrication circuits, high reliability, and many different available accessories. These pumps are renowned for their high ultimate vacuum, rapid pumping speed, quiet operation, and ability to handle vapour. The EM series has become the industry standard for laboratory and light industrial applications. Ultimate vacuum of 0.001 mbar and peak pumping speed of 12.1 CFM.