Learn More
MARSHALL SCIENTIFIC Edwards E2M28 Rotary Vane Vacuum Pump
SDP
Catalog No. NC2591260
Encompass_Preferred
The EM range of oil‑sealed rotary pumps – renowned for their high ultimate vacuum, rapid pumping speed, quiet operation and ability to handle vapour – have become the industry standard for laboratory and light industrial applications. The pump is a freestanding unit with the drive provided through a flexible coupling by either a single‑phase or three‑phase (four pole) motor.
Catalog No. NC2591260 Supplier MARSHALL SCIENTIFIC Supplier No. EDA37317984
Add to Cart
Add to Cart
Request a Quote
The EM range of oil‑sealed rotary pumps – renowned for their high ultimate vacuum, rapid pumping speed, quiet operation and ability to handle vapour – have become the industry standard for laboratory and light industrial applications. The pump is a freestanding unit with the drive provided through a flexible coupling by either a single‑phase or three‑phase (four pole) motor.