Learn More
MSC PURELL Hand Sanitizer Foam, 72 Percent Alcohol, FMX-12, 1200 mL
Catalog No. 502868038
Encompass
This advanced hand sanitizer foam is designed for effective cleaning and sanitization of hands. It comes in a convenient dispenser refill format for continuous use.
  • Foam formula for easy application
  • Capacity of 1200 mL per refill
  • Compatible with FMX-12 dispenser systems
  • Features a sanitary sealed refill to prevent contamination
  • Fragrance-free formulation
  • Meets NSF E3, Kosher, UL ECOLOGO, and USDA Certified Biobased Product standards
  • EcoLogo certified and biodegradable for environmental consideration
Catalog No. 50-286-8038 Supplier MSC Supplier No. 15824709
This advanced hand sanitizer foam is designed for effective cleaning and sanitization of hands. It comes in a convenient dispenser refill format for continuous use.
  • Foam formula for easy application
  • Capacity of 1200 mL per refill
  • Compatible with FMX-12 dispenser systems
  • Features a sanitary sealed refill to prevent contamination
  • Fragrance-free formulation
  • Meets NSF E3, Kosher, UL ECOLOGO, and USDA Certified Biobased Product standards
  • EcoLogo certified and biodegradable for environmental consideration