Learn More
Medchemexpress LLC E3 ligase ligand 32 | 2300099-98-1 | MFCD34595287 | 99.9% | 338.16 | C13H12BrN3O3 | 25 G
SDP
Catalog No. 5000285045
Encompass_Preferred
E3 ligase Ligand 32 is a small-molecule ligand for E3 ubiquitin ligases designed as a building block for PROTAC (proteolysis-targeting chimera) synthesis. The compound is provided as a solid with high analytical purity and is characterized by CAS 2300099-98-1, molecular formula C13H12BrN3O3, and molecular weight 338.16. Manufacturer documentation includes handling, storage, and purity information suitable for research use.
  • Can be used to synthesize PROTAC degraders and link to target ligands.
  • High purity suitable for research and synthesis applications.
  • Supplied as an off-white to light brown solid for ease of handling.
  • Available in multiple package sizes including gram-scale quantities.
  • Stable when stored under recommended conditions for extended periods.
  • Characterized by CAS and molecular data for unambiguous identification.
Catalog No. 50-002-85045 Supplier Medchemexpress LLC Supplier No. HYW38203825G
E3 ligase Ligand 32 is a small-molecule ligand for E3 ubiquitin ligases designed as a building block for PROTAC (proteolysis-targeting chimera) synthesis. The compound is provided as a solid with high analytical purity and is characterized by CAS 2300099-98-1, molecular formula C13H12BrN3O3, and molecular weight 338.16. Manufacturer documentation includes handling, storage, and purity information suitable for research use.
  • Can be used to synthesize PROTAC degraders and link to target ligands.
  • High purity suitable for research and synthesis applications.
  • Supplied as an off-white to light brown solid for ease of handling.
  • Available in multiple package sizes including gram-scale quantities.
  • Stable when stored under recommended conditions for extended periods.
  • Characterized by CAS and molecular data for unambiguous identification.