Learn More
Medchemexpress LLC HY-43961 100mg Medchemexpress, E3 ligase Ligand 8 CAS:1225383-33-4 Purity:>98%
SDP
Catalog No. 501932866
Encompass_Preferred
Medchemexpress, HY-43961 100mg E3 ligase Ligand 8 CAS:1225383-33-4 E3 ligase Ligand 8 is a ligand for E3 ubiquitin ligase. E3 ligase Ligand 8 can be connected to the ligand for protein by a linker to form PROTACs. PROTACs are inducers of ubiquitination-mediated degradation of cancer-promoting proteins [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2866 Supplier Medchemexpress LLC Supplier No. HY43961100MG
Medchemexpress, HY-43961 100mg E3 ligase Ligand 8 CAS:1225383-33-4 E3 ligase Ligand 8 is a ligand for E3 ubiquitin ligase. E3 ligase Ligand 8 can be connected to the ligand for protein by a linker to form PROTACs. PROTACs are inducers of ubiquitination-mediated degradation of cancer-promoting proteins [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.