Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC E3 ligase Ligand PG | 14149-34-9 | C11H11NO2 | 250 MG
SDP

Catalog No. 5000374241
Encompass_Preferred

E3 ligase Ligand PG is an E3 ligase ligand used for the recruitment of the CRBN protein. It demonstrates potent binding activity with CRBN (IC50 of 2.191 μM) and can be connected via a linker to form PROTACs, such as PROTAC PD-L1 degrader-1. This product is for research use only.

  • Used for recruitment of CRBN protein
  • Demonstrates potent binding activity with CRBN (IC50 of 2.191 μM)
  • Can be connected by a linker to form PROTACs
  • Solid, white to off-white in appearance
  • Purity of 99.94%
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • In solvent storage: -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 116.67 mg/mL (616.62 mM)

Catalog No. 50-003-74241 Supplier Medchemexpress LLC Supplier No. 250MGHY150839
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

E3 ligase Ligand PG is an E3 ligase ligand used for the recruitment of the CRBN protein. It demonstrates potent binding activity with CRBN (IC50 of 2.191 μM) and can be connected via a linker to form PROTACs, such as PROTAC PD-L1 degrader-1. This product is for research use only.

  • Used for recruitment of CRBN protein
  • Demonstrates potent binding activity with CRBN (IC50 of 2.191 μM)
  • Can be connected by a linker to form PROTACs
  • Solid, white to off-white in appearance
  • Purity of 99.94%
  • Powder storage: -20°C for 3 years or 4°C for 2 years
  • In solvent storage: -80°C for 6 months or -20°C for 1 month
  • Soluble in DMSO at 116.67 mg/mL (616.62 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.