Learn More
Cayman Chemical E3 Ligase Ligand 2, Purity: ≥95%, CAS: 5054-59-1, 5 g
SDP
Catalog No. 502909448
Encompass_Preferred
An active metabolite of thalidomide and a PROTAC synthetic intermediate; decreases production of TNF-α induced by TPA in THP-1 cells at 30 µM; completely inhibits the proliferation HUVECs at 333 µM; inhibits VEGF- and FGF2-induced angiogenesis by 14% in a CAM assay in utero at 100 µg/embryo
Catalog No. 50-290-9448 Supplier Cayman Chemical Supplier No. 366645 G
An active metabolite of thalidomide and a PROTAC synthetic intermediate; decreases production of TNF-α induced by TPA in THP-1 cells at 30 µM; completely inhibits the proliferation HUVECs at 333 µM; inhibits VEGF- and FGF2-induced angiogenesis by 14% in a CAM assay in utero at 100 µg/embryo