Learn More
Cayman Chemical E3 Ligand-Linker Conjugate 3 (hydrochloride), Purity: ≥95%, CAS: 2245697-86-1, 10 mg
SDP
Catalog No. 502913247
Encompass_Preferred
A PROTAC building block containing thalidomide; has been used in the synthesis of PROTACs for the degradation ALK and BET family of proteins with anticancer activity
Catalog No. 50-291-3247 Supplier Cayman Chemical Supplier No. 3666610 MG
A PROTAC building block containing thalidomide; has been used in the synthesis of PROTACs for the degradation ALK and BET family of proteins with anticancer activity