Learn More
Alkali Scientific E3000 UV Transilluminator
SDP
Catalog No. 503029635
Encompass_Preferred
The Alkali ScientificE3000 UV Transilluminators feature space-saving designs to fit perfectly in any busy laboratory Two models are available our standard E3000 and new E3100 myView compact model Choose your model depending on your application and the size or number of gels to analyze 302nm midrange UV wavelength Optimal for EtBr SmartGlow or SYBR stained nucleic acids E3000 models offers dual intensity settings (50% or 100%) Hinged UV safety cover Use with SmartDocenclosure for gel imaging with a smart phone The new ES3000 UV Transilluminator features a space-saving design with a width of only 10 5 inches to fit perfectly in any busy laboratory However the 16 x 19 cm UV viewing surface is sized to accommodate large gels or multiple smaller gels The illuminated surface is comprised of a special black glass which is easy to clean durable and scratch resistant An array of high intensity UV bulbs coupled with an optimized filter provides even illumination across the viewing surface
Catalog No. 50-302-9635 Supplier Alkali Scientific Supplier No. ES3000
The Alkali ScientificE3000 UV Transilluminators feature space-saving designs to fit perfectly in any busy laboratory Two models are available our standard E3000 and new E3100 myView compact model Choose your model depending on your application and the size or number of gels to analyze 302nm midrange UV wavelength Optimal for EtBr SmartGlow or SYBR stained nucleic acids E3000 models offers dual intensity settings (50% or 100%) Hinged UV safety cover Use with SmartDocenclosure for gel imaging with a smart phone The new ES3000 UV Transilluminator features a space-saving design with a width of only 10 5 inches to fit perfectly in any busy laboratory However the 16 x 19 cm UV viewing surface is sized to accommodate large gels or multiple smaller gels The illuminated surface is comprised of a special black glass which is easy to clean durable and scratch resistant An array of high intensity UV bulbs coupled with an optimized filter provides even illumination across the viewing surface