Learn More
Alkali Scientific Alkali Scientific E4100 SmartBlue™ Mini Transilluminator
SDP
Catalog No. 503029645
Encompass_Preferred
The Alkali Scientific SmartBlue E4101 Alkali Scientific SmartBlue Transilluminators have been developed for viewing DNA gels stained with new, safe fluorescent dyes.  These new dyes, such as SmartGlow, SYBR Green, Gel Green, have an excitation wavelength of 465-470nm (visible blue light), and are safer to use than Ethidium Bromide. Blue light, safe for nucleic acids Uniform illumination (UI Technology) Optimal wavelength (465nm) for common safe stains Adjustable position, amber filter covers Optional amber viewing glasses: E4000-VG1
Catalog No. 50-302-9645 Supplier Alkali Scientific Supplier No. ES4100
The Alkali Scientific SmartBlue E4101 Alkali Scientific SmartBlue Transilluminators have been developed for viewing DNA gels stained with new, safe fluorescent dyes.  These new dyes, such as SmartGlow, SYBR Green, Gel Green, have an excitation wavelength of 465-470nm (visible blue light), and are safer to use than Ethidium Bromide. Blue light, safe for nucleic acids Uniform illumination (UI Technology) Optimal wavelength (465nm) for common safe stains Adjustable position, amber filter covers Optional amber viewing glasses: E4000-VG1