Learn More
Alkali Scientific E5001-SDB SmartDoc 2.0 Gel Ima
SDP
Catalog No. 503029605
Encompass_Preferred
The Alkali Scientific E5001-SDB SmartDoc 2 0 Gel Imaging System with blue light illumination has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera Blue light is used instead of UV which is safer for lab personnel and will not damage DNA samples The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with green fluorescent dyes (SYBR Green SmartGlow GelStar GelGreen etc) The orange cover fits on top of the illumination base and acts as a filter to absorb blue light allowing visualization of the fluorescing samples The cover can also be set at an angle for gel access and cutting out DNA bands for further study Imaging enclosure works with all common smart phones System includes blue light illumination base orange viewing cover and orange photo filter Ideal for common safe stains (SmartGlow SYBR Green GelGreen etc ) Visible
Catalog No. 50-302-9605 Supplier Alkali Scientific Supplier No. RE5001SDB
The Alkali Scientific E5001-SDB SmartDoc 2 0 Gel Imaging System with blue light illumination has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera Blue light is used instead of UV which is safer for lab personnel and will not damage DNA samples The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with green fluorescent dyes (SYBR Green SmartGlow GelStar GelGreen etc) The orange cover fits on top of the illumination base and acts as a filter to absorb blue light allowing visualization of the fluorescing samples The cover can also be set at an angle for gel access and cutting out DNA bands for further study Imaging enclosure works with all common smart phones System includes blue light illumination base orange viewing cover and orange photo filter Ideal for common safe stains (SmartGlow SYBR Green GelGreen etc ) Visible