Learn More
Grainger E5405 Glove Clip With Belt Clip 1 D
Supplier:  Grainger 16N695
Encompass_Preferred
The Squids by Ergodyne Glove Holder Clip is a plastic clip that attaches to a belt or pocket to hold work gloves. It features a spring-loaded design to securely grip gloves up to 1/2" thick. The durable plastic clip measures 2-1/2" x 1-3/4" x 1" and has a black finish. This glove holder provides easy access to gloves and keeps them off dirty surfaces when not in use. It is a useful accessory for workers who need to take gloves on and off frequently. The Squids glove holder is made in the USA by Ergodyne, a leading manufacturer of work gear and safety equipment.
Catalog No. 50-276-4464