Learn More
Cayman Chemical E6 BerbamIn 10mg
SDP
Catalog No. 502982118
Encompass_Preferred
A calmodulin inhibitor that disrupts the activity of calmodulin-dependent enzymes such as myosin light chain kinase (IC50 = 8 µM) and phosphodiesterase 1A (IC50 = 0.53 µM); inhibits P-glycoprotein activity in purified primary cultured rat brain microvessel endothelial cells at 10 µM
Catalog No. 50-298-2118 Supplier Cayman Chemical Supplier No. 1935810 MG
A calmodulin inhibitor that disrupts the activity of calmodulin-dependent enzymes such as myosin light chain kinase (IC50 = 8 µM) and phosphodiesterase 1A (IC50 = 0.53 µM); inhibits P-glycoprotein activity in purified primary cultured rat brain microvessel endothelial cells at 10 µM