Learn More
TARGETMOL CHEMICALS INC E6446 5MG
SDP
Catalog No. 502579930
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg and bulk. Please contact Fisher for quotes. E6446 inhibits Toll-like receptor (TLR)7 and 9 signaling. E6446 works in a variety of human and mouse cell types and inhibits DNA-TLR9 interaction in vitro. When administered to mice this compound suppress responses to challenge doses of cytidine-phosphate-guanidine (CpG)-containing DNA which stimulates TLR9. purity: 99%
Catalog No. 50-257-9930 Supplier TARGETMOL CHEMICALS INC Supplier No. T4206
Also available in 1 mg 2 mg 10 mg 25 mg and bulk. Please contact Fisher for quotes. E6446 inhibits Toll-like receptor (TLR)7 and 9 signaling. E6446 works in a variety of human and mouse cell types and inhibits DNA-TLR9 interaction in vitro. When administered to mice this compound suppress responses to challenge doses of cytidine-phosphate-guanidine (CpG)-containing DNA which stimulates TLR9. purity: 99%