Learn More
TARGETMOL CHEMICALS INC E6446 5mg
SDP
Catalog No. NC1624951
Encompass_Preferred
E6446 inhibits Toll-like receptor (TLR)7 and 9 signaling. E6446 works in a variety of human and mouse cell types and inhibits DNA-TLR9 interaction in vitro. When administered to mice, this compound suppress responses to challenge doses of cytidine-phosphate-guanidine (CpG)-containing DNA, which stimulates TLR9.
Catalog No. NC1624951 Supplier TARGETMOL CHEMICALS INC Supplier No. T42065MG
E6446 inhibits Toll-like receptor (TLR)7 and 9 signaling. E6446 works in a variety of human and mouse cell types and inhibits DNA-TLR9 interaction in vitro. When administered to mice, this compound suppress responses to challenge doses of cytidine-phosphate-guanidine (CpG)-containing DNA, which stimulates TLR9.