Learn More
Selleck Chemical LLC E6446 dihydrochloride
SDP
Supplier:  Selleck Chemical LLC S671925MG
Encompass_Preferred
E6446 dihydrochloride is a specific Toll-like receptor 9 inhibitor
Catalog No. 50-000-01947