Learn More
Cayman Chemical E6446 (hydrochloride), Purity: ≥95%, CAS: 1345675-25-3, 10 mg
SDP
Catalog No. 502917338
Encompass_Preferred
A TLR7 and TLR9 antagonist (IC50s = 1.78 and 0.01 µM, respectively); selective for TLR7 and TLR9 over TLR4 (IC50 = 10.58 µM); inhibits CpG ODN2216- or RNA40-induced IL-6 production in isolated mouse BMDCs; prevents increases in left ventricle chamber size and cardiac fibrosis in a mouse model of transverse aortic constriction-induced heart failure at 1.5 mg/animal; increases survival and reduces increases in right ventricular systolic pressure and hypertrophy in a rat model of monocrotaline-induced pulmonary hypertension at 10 mg/kg; has been used to enhance the transfection efficiency of cell-penetrating peptides for the delivery of mRNA in vitro
Catalog No. 50-291-7338 Supplier Cayman Chemical Supplier No. 3729610 MG
A TLR7 and TLR9 antagonist (IC50s = 1.78 and 0.01 µM, respectively); selective for TLR7 and TLR9 over TLR4 (IC50 = 10.58 µM); inhibits CpG ODN2216- or RNA40-induced IL-6 production in isolated mouse BMDCs; prevents increases in left ventricle chamber size and cardiac fibrosis in a mouse model of transverse aortic constriction-induced heart failure at 1.5 mg/animal; increases survival and reduces increases in right ventricular systolic pressure and hypertrophy in a rat model of monocrotaline-induced pulmonary hypertension at 10 mg/kg; has been used to enhance the transfection efficiency of cell-penetrating peptides for the delivery of mRNA in vitro