Learn More
Medchemexpress LLC HY-122895 5mg Medchemexpress, E64FC26 CAS:2285446-62-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1228955MG
Encompass_Preferred
Medchemexpress, HY-122895 5mg E64FC26 CAS:2285446-62-8 E64FC26 is a potent pan-inhibitor of the protein disulfide isomerase (PDI) family, with IC50s of 1.9, 20.9, 25.9, 16.3, and 25.4 μM against PDIA1, PDIA3, PDIA4, TXNDC5, and PDIA6, respectively. E64FC26 shows anti-myeloma activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0997