Learn More
TARGETMOL CHEMICALS INC E7016 5MG
SDP
Catalog No. 502579541
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. E7016 (GPI 21016) is an orally available PARP inhibitor. E7016 inhibits of DNA repair. E7016 can enhance tumor cell radiosensitivity in vitro and in vivo. purity: 98%
Catalog No. 50-257-9541 Supplier TARGETMOL CHEMICALS INC Supplier No. T611895MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. E7016 (GPI 21016) is an orally available PARP inhibitor. E7016 inhibits of DNA repair. E7016 can enhance tumor cell radiosensitivity in vitro and in vivo. purity: 98%