Learn More
Selleck Chemical LLC Lenvatinib (E7080) 10mg 417716-92-8
SDP
Supplier:  Selleck Chemical LLC S116410MG
Encompass_Preferred
Lenvatinib (E7080) is a multi-target inhibitor, mostly for VEGFR2(KDR)/VEGFR3(Flt-4) with IC50 of 4 nM/5.2 nM, less potent against VEGFR1/Flt-1, ~10-fold more selective for VEGFR2/3 against FGFR1, PDGFR?/? in cell-free assays. Lenvatinib (E7080) also inhibits FGFR1-4, PDGFR, Kit (c-Kit), RET (c-RET), and shows potent antitumor activities. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-814-4