Learn More
Medchemexpress LLC E7090 succinate | 1879965-80-6 | 99.2% | 764.82 g/mol | C32H37N5O6·3/2C4H6O4 | 1 ML
SDP
Catalog No. 5000248892
Encompass_Preferred
E7090 succinate (tasurgratinib succinate) is an orally available, selective inhibitor of fibroblast growth factor receptors (FGFR1-4), provided as a 10 mM solution in DMSO (1 mL) for research use in in vitro pharmacology and preclinical studies targeting FGFR-driven signaling.
  • Selective inhibition of FGFR1-3 signaling, useful for target validation.
  • Provided as a 10 mM solution in DMSO, ready for immediate use in assays.
  • High purity suitable for biochemical and cell-based experiments.
  • Available in both solid and solution formats for experimental flexibility.
  • Molecular weight and formula documented for formulation and calculation needs.
Catalog No. 50-002-48892 Supplier Medchemexpress LLC Supplier No. HY101466A10MM 1ML
E7090 succinate (tasurgratinib succinate) is an orally available, selective inhibitor of fibroblast growth factor receptors (FGFR1-4), provided as a 10 mM solution in DMSO (1 mL) for research use in in vitro pharmacology and preclinical studies targeting FGFR-driven signaling.
  • Selective inhibition of FGFR1-3 signaling, useful for target validation.
  • Provided as a 10 mM solution in DMSO, ready for immediate use in assays.
  • High purity suitable for biochemical and cell-based experiments.
  • Available in both solid and solution formats for experimental flexibility.
  • Molecular weight and formula documented for formulation and calculation needs.