Learn More
Cayman Chemical E70K (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502919461
Encompass_Preferred
A peptide inhibitor of the CXCL8-GAG interaction; inhibits primary human neutrophil binding to HUVECs at 50 nM; decreases CXCL8-induced transendothelial migration of primary human neutrophils through HMEC-1 human microvascular endothelial cell layers at 1 to 1000 nM
Catalog No. 50-291-9461 Supplier Cayman Chemical Supplier No. 3695610 MG
A peptide inhibitor of the CXCL8-GAG interaction; inhibits primary human neutrophil binding to HUVECs at 50 nM; decreases CXCL8-induced transendothelial migration of primary human neutrophils through HMEC-1 human microvascular endothelial cell layers at 1 to 1000 nM