Learn More
Alkali Scientific E7500 QuadCount Automated Cell
SDP
Supplier:  Alkali Scientific RE7500
Encompass_Preferred
QuadCount Automated Cell Counters With count speeds at fast as 20 seconds per sample the Alkali Scientific E7500 QuadCountAutomated Cell Counter provides many advantages over the tedious and time consuming manual hemocytometry method The advanced software eliminates human bias in dead/live determinations reduces counting errors saves time and improves efficiency The Alkali Scientific E7500 QuadCountAutomated Cell Counter utilizes a unique 4-chambered disposable QuadSlide Four samples can be processed in one slide (twice the capacity of standard 2-chambered slides) decreasing consumable costs Each chamber has a hydrophilic inner surface with an engraved grid pattern High precision production and molding ensure that each QuadSlide meets the rigorous standards and dimensional tolerances required for optimal counting performance 3 Count Modes Available to analyze different sample volumes Quick Mode 0 15uL / 20 seconds (1 image frame) Normal Mode 0 9 uL / 30 seconds (6 image fram
Catalog No. 50-302-9606